missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Amino-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific Chemicals

$85.88 - $85.88

Chemical Identifiers

CAS 26215-14-5
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD03425794
InChI Key RUZXDTHZHJTTRO-UHFFFAOYSA-N
Synonym 7-amino-2h-1,4-benzoxazin-3 4h-one, 7-amino-2h-1,4-benzoxazin-3 4h one, 7-amino-4h-benzo 1,4 oxazin-3-one, 7-amino-2h-benzo b 1,4 oxazin-3 4h-one, 7-amino-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-amino, 7-amino-2,4-dihydro-1,4-benzoxazin-3-one, 7-amino-2h,4h-benzo e 1,4-oxazin-3-one, librarion l544
PubChem CID 2764182
IUPAC Name 7-amino-4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=C(C=C2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH31534MD
View Documents
Thermo Scientific Chemicals
H31534MD
250 mg
Each for $85.88
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26215-14-5
164.164
RUZXDTHZHJTTRO-UHFFFAOYSA-N
2764182
C1C(=O)NC2=C(O1)C=C(C=C2)N
C8H8N2O2
MFCD03425794
7-amino-2h-1,4-benzoxazin-3 4h-one, 7-amino-2h-1,4-benzoxazin-3 4h one, 7-amino-4h-benzo 1,4 oxazin-3-one, 7-amino-2h-benzo b 1,4 oxazin-3 4h-one, 7-amino-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-amino, 7-amino-2,4-dihydro-1,4-benzoxazin-3-one, 7-amino-2h,4h-benzo e 1,4-oxazin-3-one, librarion l544
7-amino-4H-1,4-benzoxazin-3-one

Specifications

26215-14-5
C8H8N2O2
250 mg
RUZXDTHZHJTTRO-UHFFFAOYSA-N
7-amino-4H-1,4-benzoxazin-3-one
2764182
97%
220°C to 222°C
MFCD03425794
7-amino-2h-1,4-benzoxazin-3 4h-one, 7-amino-2h-1,4-benzoxazin-3 4h one, 7-amino-4h-benzo 1,4 oxazin-3-one, 7-amino-2h-benzo b 1,4 oxazin-3 4h-one, 7-amino-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-amino, 7-amino-2,4-dihydro-1,4-benzoxazin-3-one, 7-amino-2h,4h-benzo e 1,4-oxazin-3-one, librarion l544
C1C(=O)NC2=C(O1)C=C(C=C2)N
164.164
164.17
7-Amino-2H-1,4-benzoxazin-3(4H)-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only