missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Bromo-1-tetralone 95.0+%, TCI America™

$88.24 - $264.70

Chemical Identifiers

CAS 32281-97-3
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD02179287
InChI Key YGVDCGFUUUJCDF-UHFFFAOYSA-N
Synonym 7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
PubChem CID 252731
IUPAC Name 7-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)Br)C(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B45451G
View Documents
TCI America
B45451G
1 g
Each for $88.24
 
B45455G
View Documents
TCI America
B45455G
5 g
Each for $264.70
 
Description

Description

Specifications

Chemical Identifiers

32281-97-3
225.085
YGVDCGFUUUJCDF-UHFFFAOYSA-N
252731
C1CC2=C(C=C(C=C2)Br)C(=O)C1
C10H9BrO
MFCD02179287
7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
7-bromo-3,4-dihydro-2H-naphthalen-1-one

Specifications

32281-97-3
White-Yellow
C10H9BrO
1 g
YGVDCGFUUUJCDF-UHFFFAOYSA-N
7-bromo-3,4-dihydro-2H-naphthalen-1-one
252731
≥95.0% (GC)
7-Bromo-1-tetralone
76°C
157°C
MFCD02179287
7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
C1CC2=C(C=C(C=C2)Br)C(=O)C1
225.085
225.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'