missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Fluoro-6-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6109203

 View more versions of this product

Catalog No. AAH6109203


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in medicine.

Solubility
Slightly soluble in water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions.
Specifications

Specifications

7-Fluoro-6-nitro-2H-1,4-benzoxazin-3(4H)-one
C8H5FN2O4
1 g
Slightly soluble in water.
C1C(=O)NC2=CC(=C(C=C2O1)F)[N+](=O)[O-]
212.136
212.14
103361-67-7
MFCD22056242
2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro, 7-fluoro-6-nitro-2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro-2,4-dihydro-1,4-benzoxazin-3-one, acmc-20m67v, 7-fluoro-6-nitro-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-fluoro-6-nitro, 7-fluoro-3,4-dihydro-3-oxo-6-nitro-2h-1,4-benzoxazine
MNUBUFBVOBPANC-UHFFFAOYSA-N
7-fluoro-6-nitro-4H-1,4-benzoxazin-3-one
13731297
97%

Chemical Identifiers

103361-67-7
212.136
MNUBUFBVOBPANC-UHFFFAOYSA-N
13731297
C1C(=O)NC2=CC(=C(C=C2O1)F)[N+](=O)[O-]
C8H5FN2O4
MFCD22056242
2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro, 7-fluoro-6-nitro-2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro-2,4-dihydro-1,4-benzoxazin-3-one, acmc-20m67v, 7-fluoro-6-nitro-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-fluoro-6-nitro, 7-fluoro-3,4-dihydro-3-oxo-6-nitro-2h-1,4-benzoxazine
7-fluoro-6-nitro-4H-1,4-benzoxazin-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions