missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Fluorooxindole, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6151106

 View more versions of this product

Catalog No. AAH6151106


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7-Fluorooxindole
Red to Brown
MFCD02179608
7-fluorooxindole, 7-fluoroindolin-2-one, 7-fluoro-oxindole, 7-fluoro-2-oxindole, 7-fluoroindolinone, 7-fluoro-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one, 7-fluoro-1,3-dihydro, 7-fluoro-1,3-dihydro-indol-2-one, 7-fluoro-2,3-dihydro-1h-indol-2-one, 1,3-dihydro-7-fluoro-2h-indol-2-one
C1C2=C(C(=CC=C2)F)NC1=O
151.14
151.14
Powder
71294-03-6
C8H6FNO
5 g
VMUIOEOYZHJLEZ-UHFFFAOYSA-N
7-fluoro-1,3-dihydroindol-2-one
3734371
95%

Chemical Identifiers

71294-03-6
151.14
VMUIOEOYZHJLEZ-UHFFFAOYSA-N
3734371
C1C2=C(C(=CC=C2)F)NC1=O
C8H6FNO
MFCD02179608
7-fluorooxindole, 7-fluoroindolin-2-one, 7-fluoro-oxindole, 7-fluoro-2-oxindole, 7-fluoroindolinone, 7-fluoro-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one, 7-fluoro-1,3-dihydro, 7-fluoro-1,3-dihydro-indol-2-one, 7-fluoro-2,3-dihydro-1h-indol-2-one, 1,3-dihydro-7-fluoro-2h-indol-2-one
7-fluoro-1,3-dihydroindol-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 676-237-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only