missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Hydroxyflavanone 98.0+%, TCI America™
$86.51 - $86.51
Chemical Identifiers
CAS | 6515-36-2 |
---|---|
Molecular Formula | C15H12O3 |
Molecular Weight (g/mol) | 240.258 |
MDL Number | MFCD00017487 |
InChI Key | SWAJPHCXKPCPQZ-UHFFFAOYSA-N |
Synonym | 7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo |
PubChem CID | 1890 |
ChEBI | CHEBI:34483 |
IUPAC Name | 7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one |
SMILES | C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H10061G
|
TCI America
H10061G |
1 g |
Each for $86.51
|
|
|||||
Chemical Identifiers
6515-36-2 | |
240.258 | |
SWAJPHCXKPCPQZ-UHFFFAOYSA-N | |
1890 | |
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one |
C15H12O3 | |
MFCD00017487 | |
7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo | |
CHEBI:34483 | |
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3 |
Specifications
6515-36-2 | |
C15H12O3 | |
1 g | |
SWAJPHCXKPCPQZ-UHFFFAOYSA-N | |
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one | |
1890 | |
240.26 | |
Crystalline Powder |
190°C | |
MFCD00017487 | |
7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo | |
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3 | |
240.258 | |
CHEBI:34483 | |
≥98.0% (HPLC,T) | |
7-Hydroxyflavanone |
Safety and Handling
TSCA : No