missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxyflavone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1853806

 View more versions of this product

Catalog No. AAA1853806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7-Hydroxyflavone
245°C to 247°C
MFCD00006835
194089
MQGPSCMMNJKMHQ-UHFFFAOYSA-N
7-hydroxy-2-phenylchromen-4-one
5281894
238.25
6665-86-7
C15H10O3
5 g
7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O
238.242
CHEBI:2268
98%

Chemical Identifiers

6665-86-7
238.242
MQGPSCMMNJKMHQ-UHFFFAOYSA-N
5281894
7-hydroxy-2-phenylchromen-4-one
C15H10O3
MFCD00006835
7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11
CHEBI:2268
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 229-705-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only