missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Hydroxyflavone 97.0+%, TCI America™
Supplier: TCI America H08521G
Specifications
7-Hydroxyflavone | |
246°C | |
C15H10O3 | |
1 g | |
MQGPSCMMNJKMHQ-UHFFFAOYSA-N | |
7-hydroxy-2-phenylchromen-4-one | |
5281894 | |
238.24 | |
Crystalline Powder |
6665-86-7 | |
Yellow | |
MFCD00006835 | |
7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11 | |
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O | |
238.242 | |
CHEBI:2268 | |
≥97.0% (T) |
Chemical Identifiers
6665-86-7 | |
238.242 | |
MQGPSCMMNJKMHQ-UHFFFAOYSA-N | |
5281894 | |
7-hydroxy-2-phenylchromen-4-one |
C15H10O3 | |
MFCD00006835 | |
7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11 | |
CHEBI:2268 | |
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O |
Safety and Handling
TSCA : No