Learn More
7-Methoxy-1-tetralone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 172430250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
7-Methoxy-1-tetralone | |
96.0 | |
61°C to 63°C | |
Authentic | |
Glass bottle | |
MFCD00001696 | |
09, II, 889 | |
GABLTKRIYDNDIN-UHFFFAOYSA-N | |
7-methoxy-3,4-dihydro-2H-naphthalen-1-one | |
81276 | |
97% |
6836-19-7 | |
100.0 | |
White to Beige | |
96% min. (GC) | |
C11H12O2 | |
25 g | |
7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone | |
COC1=CC2=C(CCCC2=O)C=C1 | |
176.21 | |
176.21 | |
Crystalline Powder |
Chemical Identifiers
6836-19-7 | |
176.21 | |
GABLTKRIYDNDIN-UHFFFAOYSA-N | |
81276 | |
COC1=CC2=C(CCCC2=O)C=C1 |
C11H12O2 | |
MFCD00001696 | |
7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone | |
7-methoxy-3,4-dihydro-2H-naphthalen-1-one |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 229-916-
RUO – Research Use Only