missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methoxycoumarin-4-acetic acid, Thermo Scientific Chemicals

Precursor to coumarin-type fluorescent reagents

Supplier:  Thermo Scientific Chemicals J62475MC

 View more versions of this product

Catalog No. AAJ62475MC


Explore available promotions

Description

Description

7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

Solubility
Soluble in acetone and dimethyl sulfoxide.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
Specifications

Specifications

7-Methoxycoumarin-4-acetic acid
193°C
87°C (189°F)
MFCD00009774
7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
ZEKAXIFHLIITGV-UHFFFAOYSA-N
2-(7-methoxy-2-oxochromen-4-yl)acetic acid
342221
234.21
62935-72-2
Green
C12H10O5
100 mg
Soluble in acetone and dimethyl sulfoxide.
COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1
234.21
CHEBI:51666
Crystalline Solid

Chemical Identifiers

62935-72-2
234.21
ZEKAXIFHLIITGV-UHFFFAOYSA-N
342221
2-(7-methoxy-2-oxochromen-4-yl)acetic acid
C12H10O5
MFCD00009774
7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
CHEBI:51666
COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only