missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Phenyl-1-heptanol, 97%, Thermo Scientific Chemicals

$158.33 - $620.90

Chemical Identifiers

CAS 3208-25-1
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00037315
InChI Key UXMUSYTXSNVRMW-UHFFFAOYSA-N
Synonym benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
PubChem CID 154271
IUPAC Name 7-phenylheptan-1-ol
SMILES C1=CC=C(C=C1)CCCCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1253303
View Documents
Thermo Scientific Chemicals
L1253303
1 g
Each for $158.33
 
AAL1253306
View Documents
Thermo Scientific Chemicals
L1253306
5 g
Each for $620.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3208-25-1
192.302
UXMUSYTXSNVRMW-UHFFFAOYSA-N
154271
C1=CC=C(C=C1)CCCCCCCO
C13H20O
MFCD00037315
benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
7-phenylheptan-1-ol

Specifications

3208-25-1
115°C to 117°C (0.5 mmHg)
1.508
1 g
benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
C1=CC=C(C=C1)CCCCCCCO
192.302
192.3
7-Phenyl-1-heptanol
0.962
C13H20O
MFCD00037315
1865431
UXMUSYTXSNVRMW-UHFFFAOYSA-N
7-phenylheptan-1-ol
154271
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only