Moldex™ 7800 Premium Silicone Half Mask
Moldex 7800 Series Reusable Silicone Half Mask Respirator is specifically designed to comfortably conform to facial contours and with a modern look that increases worker acceptance. The curved neck buckle and adjustable head cradle ensure a comfortable, custom fit for most users.
Supplier: Moldex™ 7802
Description
- Soft Silicone facepiece feels supple on the face, yet firmly retains its shape.
- Sleek design for modern, high-tech look and increased worker acceptance.
- Light weight design plus extra-wide flexible face seal delivers all-day comfort.
- Low profile, compact design provides wide field of vision and fits well under welding helmets and with safety glasses.
- Mask drops down for convenient storage around the neck or locks down for custom fit.
- Adjustable head cradle and curved neck buckles for extra comfort.
- Maintenance made simple with very few replacement parts.
Specifications
Black | |
Silicone | |
Medium | |
Yes |
Reusable | |
Reusable Respirator | |
No | |
12/Cs. |