MP Biomedicals™ 7X™-O-Matic Solution
7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)
Supplier: MP Biomedicals™ 097667493
Description
- Developed for safe and effective cleaning of laboratory instruments and glassware
- Formulated to be used in a machine washer to withstand heat, pressure and agitation
- Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
- Behaves and remains mostly suds-free when agitated
- Several convenient package sizes are available, and it is stable for long-term storage
- Effective and safe
- Eliminates residue leaving no interfering fluorescence
- No etch to glass or plastic labware at any concentration
- Completely drains from glassware in seconds
Specifications
Liquid | |
Clear to Light Yellow | |
15°C to 30°C | |
1 gal. |
9 to 11 | |
7X | |
Use on Glass, Plastic, benchtop equipment and tubing |