MP Biomedicals™ 7X™-O-Matic Solution
7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)
Supplier:  MP Biomedicals™ 097667494
 View more versions of this product
Catalog No. ICN7667494
Explore available promotions
Description
Description
  • Developed for safe and effective cleaning of laboratory instruments and glassware
  • Formulated to be used in a machine washer to withstand heat, pressure and agitation
  • Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available, and it is stable for long-term storage
  • Effective and safe
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
Specifications
Specifications
Liquid
Clear to Light Yellow
15°C to 30°C
4 Pk.
9 to 11
7X
Use on Glass, Plastic, benchtop equipment and tubing
1 gal.