missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Acetyl-7-hydroxy-4-methylcoumarin 98.0+%, TCI America™
Supplier: TCI America A18485G
Specifications
8-Acetyl-7-hydroxy-4-methylcoumarin | |
170°C | |
C12H10O4 | |
5 g | |
WZOMQVFUPMLOGT-UHFFFAOYSA-N | |
8-acetyl-7-hydroxy-4-methyl-2H-chromen-2-one | |
5324648 | |
≥98.0% (GC) |
2555-29-5 | |
Yellow | |
MFCD00100602 | |
8-Acetyl-4-methylumbelliferone | |
CC(=O)C1=C2OC(=O)C=C(C)C2=CC=C1O | |
218.21 | |
218.21 | |
Crystalline Powder |
Chemical Identifiers
2555-29-5 | |
218.21 | |
WZOMQVFUPMLOGT-UHFFFAOYSA-N | |
5324648 | |
CC(=O)C1=C2OC(=O)C=C(C)C2=CC=C1O |
C12H10O4 | |
MFCD00100602 | |
8-Acetyl-4-methylumbelliferone | |
8-acetyl-7-hydroxy-4-methyl-2H-chromen-2-one |
Safety and Handling
RTECSNumber : GN4910000
TSCA : No