missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1675214

 View more versions of this product

Catalog No. AAA1675214


Explore available promotions

Description

Description

8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.
Specifications

Specifications

8-Aminoquinoline
63°C to 67°C
C9H8N2
25 g
Air Sensitive
Soluble in water.
C1=CC2=C(C(=C1)N)N=CC=C2
144.177
144.18
578-66-5
174°C (26 mmHg)
MFCD00006809
114474
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
WREVVZMUNPAPOV-UHFFFAOYSA-N
quinolin-8-amine
11359
≥98%

Chemical Identifiers

578-66-5
144.177
WREVVZMUNPAPOV-UHFFFAOYSA-N
11359
C1=CC2=C(C(=C1)N)N=CC=C2
C9H8N2
MFCD00006809
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
quinolin-8-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H341

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 209-427-9

RTECSNumber : VA9627000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only