missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Bromo-1-naphthoic acid, Technical Grade, Maybridge™
Supplier: Thermo Scientific Chemicals BTB12326DA
Specifications
8-Bromo-1-naphthoic acid | |
95% | |
C11H7BrO2 | |
8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid | |
C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br | |
251.079 | |
251.08 |
1729-99-3 | |
Amber Glass Bottle | |
1 g | |
DMEZDDHJCUHENA-UHFFFAOYSA-N | |
8-bromonaphthalene-1-carboxylic acid | |
72873 | |
95% |
Chemical Identifiers
1729-99-3 | |
251.079 | |
8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid | |
8-bromonaphthalene-1-carboxylic acid |
C11H7BrO2 | |
DMEZDDHJCUHENA-UHFFFAOYSA-N | |
72873 | |
C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br |