missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98.0+%, TCI America™

$173.37 - $173.37

Chemical Identifiers

CAS 666816-98-4
Molecular Formula C10H9BrN4O2
Molecular Weight (g/mol) 297.112
MDL Number MFCD22126091
InChI Key HFZOBQSHTNNKFY-UHFFFAOYSA-N
Synonym 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
PubChem CID 21088927
IUPAC Name 8-bromo-7-but-2-ynyl-3-methylpurine-2,6-dione
SMILES CC#CCN1C2=C(N=C1Br)N(C(=O)NC2=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4454200MG
View Documents
TCI America
B4454200MG
200 mg
Each for $173.37
Only null left
 
Description

Description

Specifications

Chemical Identifiers

666816-98-4
297.112
HFZOBQSHTNNKFY-UHFFFAOYSA-N
21088927
CC#CCN1C2=C(N=C1Br)N(C(=O)NC2=O)C
C10H9BrN4O2
MFCD22126091
8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
8-bromo-7-but-2-ynyl-3-methylpurine-2,6-dione

Specifications

666816-98-4
White
MFCD22126091
8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
CC#CCN1C2=C(N=C1Br)N(C(=O)NC2=O)C
297.112
297.11
Crystalline Powder
285°C
C10H9BrN4O2
200 mg
HFZOBQSHTNNKFY-UHFFFAOYSA-N
8-bromo-7-but-2-ynyl-3-methylpurine-2,6-dione
21088927
≥98.0% (HPLC,T)
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'