missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Chloro-1-octanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H28417AC

 View more versions of this product

Catalog No. AAH28417AC


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

8-Chloro-1-octanol
0.976
>110°C (230°F)
1.458
25 mL
YDFAJMDFCCJZSI-UHFFFAOYSA-N
8-chlorooctan-1-ol
90012
98%
23144-52-7
129°C to 130°C (11 mmHg)
C8H17ClO
MFCD00040005
8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
C(CCCCCl)CCCO
164.673
164.67

Chemical Identifiers

23144-52-7
164.673
YDFAJMDFCCJZSI-UHFFFAOYSA-N
90012
C(CCCCCl)CCCO
C8H17ClO
MFCD00040005
8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
8-chlorooctan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 245-451-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only