missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Chlorotheophylline 98.0+%, TCI America™
Supplier: TCI America C0293250G
Specifications
8-Chlorotheophylline | |
White-Yellow | |
MFCD00005581 | |
8-chlorotheophylline, 1,3-dimethyl-8-chloroxanthine, theophylline, 8-chloro, 8-chloro-theophylline, unii-ge2ua340fm, 8-chloro-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-chloro-3,7-dihydro-1,3-dimethyl, 8-chloro-3,7-dihydro-1,3-dimethyl-1h-purine-2,6-dione, ge2ua340fm, chembl88611 | |
CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O | |
214.61 | |
CHEBI:59771 | |
≥98.0% (HPLC,T) |
85-18-7 | |
C7H7ClN4O2 | |
250 g | |
RYIGNEOBDRVTHA-UHFFFAOYSA-N | |
8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione | |
10661 | |
214.61 | |
Crystalline Powder |
Chemical Identifiers
85-18-7 | |
214.61 | |
RYIGNEOBDRVTHA-UHFFFAOYSA-N | |
10661 | |
8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione |
C7H7ClN4O2 | |
MFCD00005581 | |
8-chlorotheophylline, 1,3-dimethyl-8-chloroxanthine, theophylline, 8-chloro, 8-chloro-theophylline, unii-ge2ua340fm, 8-chloro-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-chloro-3,7-dihydro-1,3-dimethyl, 8-chloro-3,7-dihydro-1,3-dimethyl-1h-purine-2,6-dione, ge2ua340fm, chembl88611 | |
CHEBI:59771 | |
CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O |
Safety and Handling
EINECSNumber : (9)-0379
RTECSNumber : XH5063000
TSCA : No