missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Code Corporation™ 8 ft. Coiled RS232 Cable
Use this 8 ft. Coiled RS232 Affinity™ Cable CRA-C511 with Code™'s cabled barcode readers.
Supplier: Code Corporation™ 176700
Description
- Reader termination: 10-pin RJ45
- Host termination: 9-pin, female, D-Sub connector
Specifications
| Code Readers | |
| Cable | |
| 9 pin D-Sub | |
| CRA-C501 |