missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 98%

CAS: 32178-63-5 | C12H14O3 | 206.241 g/mol

$99.90 - $185.36

Chemical Identifiers

CAS 32178-63-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01075657
InChI Key VOAFGTBETRJDPU-UHFFFAOYSA-N
Synonym 8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
PubChem CID 3624709
IUPAC Name 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
SMILES COC1=CC=CC2=C1CC(CC2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL13751MD
View Documents
Thermo Scientific Chemicals
L13751MD
250 mg N/A
Only null left
 
AAL1375103
View Documents
Thermo Scientific Chemicals
L1375103
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32178-63-5
206.241
VOAFGTBETRJDPU-UHFFFAOYSA-N
3624709
COC1=CC=CC2=C1CC(CC2)C(=O)O
C12H14O3
MFCD01075657
8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid

Specifications

32178-63-5
C12H14O3
250 mg
8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
COC1=CC=CC2=C1CC(CC2)C(=O)O
206.241
206.24
8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
141°C to 143°C
MFCD01075657
2269890
VOAFGTBETRJDPU-UHFFFAOYSA-N
8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
3624709
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only