missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-(p-Toluenesulfonylamino)quinoline, 97%, Thermo Scientific™
$42.21 - $525.89
Chemical Identifiers
CAS | 10304-39-9 |
---|---|
Molecular Formula | C16H14N2O2S |
Molecular Weight (g/mol) | 298.36 |
MDL Number | MFCD00168990 |
InChI Key | ZSMKPYXVUIWTCT-UHFFFAOYSA-N |
Synonym | 8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide |
PubChem CID | 82533 |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5538603
|
Thermo Scientific Chemicals
H5538603 |
1 g |
Each for $42.21
|
|
|||||
AAH5538606
|
Thermo Scientific Chemicals
H5538606 |
5 g |
Each for $134.00
|
|
|||||
AAH5538614
|
Thermo Scientific Chemicals
H5538614 |
25 g |
Each for $525.89
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10304-39-9 | |
298.36 | |
ZSMKPYXVUIWTCT-UHFFFAOYSA-N | |
82533 |
C16H14N2O2S | |
MFCD00168990 | |
8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide | |
CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1 |
Specifications
10304-39-9 | |
Brown to White | |
MFCD00168990 | |
264159 | |
ZSMKPYXVUIWTCT-UHFFFAOYSA-N | |
4-methyl-N-(quinolin-8-yl)benzene-1-sulfonamide | |
82533 | |
97% | |
8-(p-Toluenesulfonylamino)quinoline |
154°C to 158°C | |
C16H14N2O2S | |
1 g | |
8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide | |
CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1 | |
298.36 | |
298.36 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : DB2948850
TSCA : Yes
Recommended Storage : Ambient temperatures