Learn More
Agilent BioTek 800 TS Absorbance Reader
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
$10063.55 - $13278.81
Specifications
Product Type | Microplate Reader |
---|---|
Detector Type | Photodiode |
Interface | USB Type B, USB Type A |
Light Source | Tungsten Halogen Lamp |
Reading Options | Endpoint, Kinetic, Well-area Scanning |
Catalog Number | Mfr. No. | Model | Filters | Includes | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Filters | Includes | Price | Quantity | |||
BT800TS
|
Agilent BioTek
800TSSN |
800TS | 405, 450, 490, 630 nm | 6- to 96-well plates; 400 to 750 nm; Shaking |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT800TSI
|
Agilent BioTek
800TSISN |
800TSI | 405, 450, 490, 630 nm | 6- to 96-well plates; 400 to 750 nm; Shaking; Incubation |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT800TSNB
|
Agilent BioTek
800TSNBSN |
800TSNB | 405, 450, 490, 630 nm | 6- to 384-well Terasaki plates; 400 to 750 nm |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT800TSUV
|
Agilent BioTek
800TSUVSN |
800TSUV | 340, 405, 450, 490, 630 nm | 6- to 96-well plates; 340 to 750 nm; Shaking |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT800TSUVI
|
Agilent BioTek
800TSUVISN |
800TSUVI | 340, 405, 450, 490, 630 nm | 6- to 96-well plates; 340 to 750 nm; Shaking; Incubation |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Warranty
12 months
Specifications
Microplate Reader | |
USB Type B, USB Type A | |
Endpoint, Kinetic, Well-area Scanning | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
CE and TUV marked. RoHS compliant. | |
100/240 V | |
8 kg |
Photodiode | |
Tungsten Halogen Lamp | |
Linear (except NB configurations) | |
10 nm | |
Measuring absorbance in microplate-based assays | |
18.5 lb. |