Learn More
9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1571406
Description
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.
Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
9,10-Diphenylanthracene | |
248°C to 254°C | |
C26H18 | |
5 g | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
9,10-diphenylanthracene | |
15159 | |
330.43 |
1499-10-1 | |
Odorless | |
MFCD00001253 | |
1914010 | |
Soluble in ethanol,chloroform,benzene and carbon disulfide. Insoluble in water. | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 | |
330.43 | |
CHEBI:51676 | |
99% |
Chemical Identifiers
1499-10-1 | |
330.43 | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
15159 | |
9,10-diphenylanthracene |
C26H18 | |
MFCD00001253 | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
CHEBI:51676 | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
Safety and Handling
EINECSNumber : 216-105-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only