missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(4-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™
$124.89 - $628.19
Chemical Identifiers
| CAS | 894791-46-9 |
|---|---|
| Molecular Formula | C24H16BrN |
| Molecular Weight (g/mol) | 398.303 |
| MDL Number | MFCD16038226 |
| InChI Key | MOCNGNGLTRMQQH-UHFFFAOYSA-N |
| PubChem CID | 53401105 |
| IUPAC Name | 3-bromo-9-(4-phenylphenyl)carbazole |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Chemical Identifiers
| 894791-46-9 | |
| 398.303 | |
| MOCNGNGLTRMQQH-UHFFFAOYSA-N | |
| 3-bromo-9-(4-phenylphenyl)carbazole |
| C24H16BrN | |
| MFCD16038226 | |
| 53401105 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Specifications
| 894791-46-9 | |
| White-Yellow | |
| MFCD16038226 | |
| MOCNGNGLTRMQQH-UHFFFAOYSA-N | |
| 3-bromo-9-(4-phenylphenyl)carbazole | |
| 53401105 | |
| ≥98.0% (GC) | |
| 9-(4-Biphenylyl)-3-bromocarbazole |
| 155°C | |
| C24H16BrN | |
| 5 g | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 | |
| 398.303 | |
| 398.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No