missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene 98.0+%, TCI America™
$59.65 - $592.76
Chemical Identifiers
| CAS | 88938-12-9 |
|---|---|
| Molecular Formula | C27H22O2 |
| Molecular Weight (g/mol) | 378.47 |
| MDL Number | MFCD04038046 |
| InChI Key | NUDSREQIJYWLRA-UHFFFAOYSA-N |
| PubChem CID | 11014345 |
| IUPAC Name | 4-[9-(4-hydroxy-3-methylphenyl)-9H-fluoren-9-yl]-2-methylphenol |
| SMILES | CC1=C(O)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(O)C=C1 |
Chemical Identifiers
| 88938-12-9 | |
| 378.47 | |
| NUDSREQIJYWLRA-UHFFFAOYSA-N | |
| 4-[9-(4-hydroxy-3-methylphenyl)-9H-fluoren-9-yl]-2-methylphenol |
| C27H22O2 | |
| MFCD04038046 | |
| 11014345 | |
| CC1=C(O)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(O)C=C1 |
Specifications
| 88938-12-9 | |
| White | |
| MFCD04038046 | |
| 4,4′C-(9-Fluorenylidene)di(o-cresol) | |
| CC1=C(O)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(O)C=C1 | |
| 378.47 | |
| 378.47 | |
| Crystalline Powder |
| 219°C | |
| C27H22O2 | |
| 25 g | |
| NUDSREQIJYWLRA-UHFFFAOYSA-N | |
| 4-[9-(4-hydroxy-3-methylphenyl)-9H-fluoren-9-yl]-2-methylphenol | |
| 11014345 | |
| ≥98.0% (GC) | |
| 9,9-Bis(4-hydroxy-3-methylphenyl)fluorene |
Safety and Handling
EINECSNumber : (4)-1869
TSCA : No