missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-hydroxyphenyl)fluorene 96.0+%, TCI America™
$141.67 - $997.27
Chemical Identifiers
| CAS | 3236-71-3 |
|---|---|
| Molecular Formula | C25H18O2 |
| Molecular Weight (g/mol) | 350.42 |
| MDL Number | MFCD00191392 |
| InChI Key | YWFPGFJLYRKYJZ-UHFFFAOYSA-N |
| PubChem CID | 76716 |
| IUPAC Name | 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol |
| SMILES | OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 3236-71-3 | |
| 350.42 | |
| YWFPGFJLYRKYJZ-UHFFFAOYSA-N | |
| 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol |
| C25H18O2 | |
| MFCD00191392 | |
| 76716 | |
| OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Specifications
| 3236-71-3 | |
| White | |
| MFCD00191392 | |
| 3077 | |
| YWFPGFJLYRKYJZ-UHFFFAOYSA-N | |
| 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol | |
| 76716 | |
| ≥96.0% (GC) | |
| 9,9-Bis(4-hydroxyphenyl)fluorene |
| 227°C | |
| C25H18O2 | |
| 25 g | |
| 4,4′C-(9-Fluorenylidene)diphenol | |
| OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 350.42 | |
| 350.42 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1620
TSCA : Yes