missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
$216.10 - $216.10
Chemical Identifiers
CAS | 258865-48-4 |
---|---|
Molecular Formula | C29H44B2O4 |
Molecular Weight (g/mol) | 478.29 |
MDL Number | MFCD03701608 |
InChI Key | HURJMQMZDPOUOU-UHFFFAOYSA-N |
Synonym | 9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid |
PubChem CID | 4283240 |
IUPAC Name | (7-borono-9,9-dioctylfluoren-2-yl)boronic acid |
SMILES | CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5537303
|
Thermo Scientific Chemicals
H5537303 |
1 g |
Each for $216.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
258865-48-4 | |
478.29 | |
HURJMQMZDPOUOU-UHFFFAOYSA-N | |
4283240 | |
CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O |
C29H44B2O4 | |
MFCD03701608 | |
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid | |
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid |
Specifications
258865-48-4 | |
MFCD03701608 | |
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid | |
CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O | |
478.29 | |
478.29 | |
Solid |
C29H44B2O4 | |
1 g | |
HURJMQMZDPOUOU-UHFFFAOYSA-N | |
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid | |
4283240 | |
97% | |
9,9-Di-n-octylfluorene-2,7-diboronic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only