missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dioctylfluorene-2,7-diboronic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 258865-48-4
Molecular Formula C29H44B2O4
Molecular Weight (g/mol) 478.33
MDL Number MFCD03701608
InChI Key HURJMQMZDPOUOU-UHFFFAOYSA-N
Synonym 9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
PubChem CID 4283240
IUPAC Name (7-borono-9,9-dioctylfluoren-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C3=C(C2(CCCCCCCC)CCCCCCCC)C=C(C=C3)B(O)O)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398310050
SDS View Documents
Thermo Scientific Chemicals
398310050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

258865-48-4
478.33
HURJMQMZDPOUOU-UHFFFAOYSA-N
4283240
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCCCC)CCCCCCCC)C=C(C=C3)B(O)O)(O)O
C29H44B2O4
MFCD03701608
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid

Specifications

258865-48-4
100.0
97.5% min. (HPLC)
C29H44B2O4
5g
HURJMQMZDPOUOU-UHFFFAOYSA-N
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid
4283240
98%
97.5
Authentic
Glass bottle
MFCD03701608
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCCCC)CCCCCCCC)C=C(C=C3)B(O)O)(O)O
478.33
478.33
9, 9-Dioctylfluorene-2, 7-diboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only