missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$91.20 - $459.94
Specifications
Chemical Name or Material | 9-Borabicyclo[3.3.1]nonane |
---|---|
CAS | 280-64-8,109-99-9 |
CAS Min % | 6.14,92.49 |
CAS Max % | 7.51,93.86 |
Density | 0.8840g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC165811000
|
Thermo Scientific Chemicals
165811000 |
100 mL | AcroSeal™ Glass bottle |
Each for $91.20
|
|
||||
AC165818000
|
Thermo Scientific Chemicals
165818000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $459.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
9-Borabicyclo[3.3.1]nonane | |
6.14,92.49 | |
0.8840g/mL | |
C8H15B | |
02,31; 03,24; 04,41; 05,46; 06,62; 07,29; 08,47; 09,57; 10,48; 11,68; 13,92; 14,52; 15,43; 17,49; 01,732; 04,356 | |
9-borabicyclo 3.3.1 nonane, 9-borabicyclo 3.3.1 nonan, 9-borabicyclo 3,3,1 nonane, 9$l^ 2-borabicyclo 3.3.1 nonane, 1s,5s-9-borabicyclo 3.3.1 nonane, monomer; 21205-91-4 dimer, 9-borabicyclo 3.3.1 nonane,9-bbn,intetrahydrofuran | |
AMKGKYQBASDDJB-UHFFFAOYSA-N | |
9$l^{2}-borabicyclo[3.3.1]nonane | |
6327450 |
280-64-8,109-99-9 | |
7.51,93.86 | |
−17°C | |
MFCD00074742 | |
0.884 | |
Solubility in water: reacts | |
[B]1C2CCCC1CCC2 | |
122.02 | |
122.02 |
RUO – Research Use Only