Learn More
9-Borabicyclo[3.3.1]nonyl trifluoromethanesulfonate, 0.5M solution in hexanes, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 224781000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
9-Borabicyclo[3.3.1]nonyl trifluoromethanesulfonate | |
14.74,77.89 | |
0.7330g/mL | |
AcroSeal™ Glass Bottle | |
MFCD00167981 | |
0.733 | |
Solubility in water: reacts | |
B1(C2CCCC1CCC2)OS(=O)(=O)C(F)(F)F | |
270.08 | |
270.08 |
62731-43-5,110-54-3 | |
22.11,85.26 | |
−22°C | |
C9H14BF3O3S | |
100mL | |
9-bbn triflate solution, 9-borabicyclo 3.3.1 nonyl trifluoromethanesulfonate, 9-borabicyclo 3.3.1 nonan-9-yl trifluoromethanesulfonate, 9-bbntriflate, 9-bbnotf, 9-bbn triflate, pubchem18332, 9-trifluoromethylsulfonyloxy-9-borabicyclo 3.3.1 nonane, 9-bbn triflate solution, 0.5 m in hexanes, 9-bbn inverted exclamation mark currencycf3so3 | |
NXHUKDCGEGEZCX-UHFFFAOYSA-N | |
9-borabicyclo[3.3.1]nonan-9-yl trifluoromethanesulfonate | |
5222845 |
Chemical Identifiers
62731-43-5 | |
270.08 | |
NXHUKDCGEGEZCX-UHFFFAOYSA-N | |
5222845 | |
B1(C2CCCC1CCC2)OS(=O)(=O)C(F)(F)F |
C9H14BF3O3S | |
MFCD00167981 | |
9-bbn triflate solution, 9-borabicyclo 3.3.1 nonyl trifluoromethanesulfonate, 9-borabicyclo 3.3.1 nonan-9-yl trifluoromethanesulfonate, 9-bbntriflate, 9-bbnotf, 9-bbn triflate, pubchem18332, 9-trifluoromethylsulfonyloxy-9-borabicyclo 3.3.1 nonane, 9-bbn triflate solution, 0.5 m in hexanes, 9-bbn inverted exclamation mark currencycf3so3 | |
9-borabicyclo[3.3.1]nonan-9-yl trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
In contact with wa
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO â Research Use Only