missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Bromo-10-(1-naphthyl)anthracene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6402403

 View more versions of this product

Catalog No. AAH6402403


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

9-Bromo-10-(1-naphthyl)anthracene
Yellow
MFCD11046571
20386637
SYACRXBYRNYMLN-UHFFFAOYSA-N
9-bromo-10-naphthalen-1-ylanthracene
21076365
98%
400607-04-7
C24H15Br
1 g
9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
383.288
383.29
Solid

Chemical Identifiers

400607-04-7
383.288
SYACRXBYRNYMLN-UHFFFAOYSA-N
21076365
C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
C24H15Br
MFCD11046571
9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
9-bromo-10-naphthalen-1-ylanthracene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions