missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-Bromo-10-(9-phenanthryl)anthracene 98.0+%, TCI America™
Supplier: TCI America B4572200MG
Specifications
9-Bromo-10-(9-phenanthryl)anthracene | |
232°C | |
MFCD27939628 | |
OBIXPXSRUDGFLE-UHFFFAOYSA-N | |
9-bromo-10-(phenanthren-9-yl)anthracene | |
86031959 | |
≥98.0% (HPLC) |
845457-53-6 | |
C28H17Br | |
200 mg | |
BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12 | |
433.35 | |
433.35 | |
Crystalline Powder |
Chemical Identifiers
845457-53-6 | |
433.35 | |
OBIXPXSRUDGFLE-UHFFFAOYSA-N | |
9-bromo-10-(phenanthren-9-yl)anthracene |
C28H17Br | |
MFCD27939628 | |
86031959 | |
BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
TSCA : No