Learn More
9-Bromo-10-phenylanthracene, 98%, Thermo Scientific Chemicals
$119.38 - $458.55
Chemical Identifiers
CAS | 23674-20-6 |
---|---|
Molecular Formula | C20H13Br |
Molecular Weight (g/mol) | 333.228 |
MDL Number | MFCD00230983 |
InChI Key | WHGGVVHVBFMGSG-UHFFFAOYSA-N |
Synonym | 10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene |
PubChem CID | 4155836 |
IUPAC Name | 9-bromo-10-phenylanthracene |
SMILES | C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6479403
|
Thermo Scientific Chemicals
H6479403 |
1 g |
Each for $119.38
|
|
|||||
AAH6479406
|
Thermo Scientific Chemicals
H6479406 |
5 g |
Each for $458.55
|
|
|||||
Description
It is used as an OLED material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an OLED material.
Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
23674-20-6 | |
333.228 | |
WHGGVVHVBFMGSG-UHFFFAOYSA-N | |
4155836 | |
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br |
C20H13Br | |
MFCD00230983 | |
10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene | |
9-bromo-10-phenylanthracene |
Specifications
23674-20-6 | |
Yellow | |
MFCD00230983 | |
2277064 | |
Insoluble in water. Soluble in hot toluene,dimethylformamide. | |
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br | |
333.228 | |
333.23 | |
Powder |
154°C to 158°C | |
C20H13Br | |
1 g | |
10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene | |
WHGGVVHVBFMGSG-UHFFFAOYSA-N | |
9-bromo-10-phenylanthracene | |
4155836 | |
98% | |
9-Bromo-10-phenylanthracene |
Safety and Handling
GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only