missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Chloroanthracene, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0109206

 View more versions of this product

Catalog No. AAL0109206


Explore available promotions

Description

Description

9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

9-Chloroanthracene
101°C to 106°C
MFCD00001245
1869540
Insoluble in water.
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Cl
212.676
212.68
716-53-0
C14H9Cl
5 g
anthracene, 9-chloro, ccris 5547, 9-chloro-anthracene, #, anthracene,9-chloro, bidd:gt0730
KULLJOPUZUWTMF-UHFFFAOYSA-N
9-chloroanthracene
69744
96%

Chemical Identifiers

716-53-0
212.676
KULLJOPUZUWTMF-UHFFFAOYSA-N
9-chloroanthracene
C14H9Cl
MFCD00001245
69744
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H341

EINECSNumber : 211-937-1

RTECSNumber : CA9561000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only