missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Fluorenone, 98+%, Thermo Scientific Chemicals

$102.33 - $1302.85

Chemical Identifiers

CAS 486-25-9
Molecular Formula C13H8O
Molecular Weight (g/mol) 180.206
MDL Number MFCD00001141
InChI Key YLQWCDOCJODRMT-UHFFFAOYSA-N
Synonym 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
PubChem CID 10241
ChEBI CHEBI:17922
IUPAC Name fluoren-9-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1291022
View Documents
Thermo Scientific Chemicals
A1291022
100 g
Each for $102.33
 
AAA1291036
View Documents
Thermo Scientific Chemicals
A1291036
500 g
Each for $341.33
 
AAA129100E
View Documents
Thermo Scientific Chemicals
A129100E
2500 g
Each for $1,302.85
 
Description

Description

9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

Solubility
Soluble in water, alcohol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

486-25-9
180.206
YLQWCDOCJODRMT-UHFFFAOYSA-N
10241
fluoren-9-one
C13H8O
MFCD00001141
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
CHEBI:17922
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

Specifications

486-25-9
0.9
163°C (325°F)
C13H8O
100 g
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
YLQWCDOCJODRMT-UHFFFAOYSA-N
fluoren-9-one
10241
180.21
9-Fluorenone
82°C to 85°C
342°C
98%
MFCD00001141
1636531
Soluble in water,alcohol,acetone,ether and benzene.
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
180.206
CHEBI:17922
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-630-7

RTECSNumber : LL8925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only