missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Fluorenylmethyl chloroformate, 98+%, Thermo Scientific Chemicals

Store cold

Supplier:  Thermo Scientific Chemicals A1168303

 View more versions of this product

Catalog No. AAA1168303


Explore available promotions

Description

Description

Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.

Solubility
Soluble in dioxane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols and amines.
Specifications

Specifications

9-Fluorenylmethyl chloroformate
60°C to 64°C
C15H11ClO2
1 g
2279177
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
IRXSLJNXXZKURP-UHFFFAOYSA-N
9H-fluoren-9-ylmethyl carbonochloridate
34367
98%
28920-43-6
98%
MFCD00001138
UN2923
Moisture sensitive
Soluble in dioxane.
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl
258.701
258.7

Chemical Identifiers

28920-43-6
258.701
IRXSLJNXXZKURP-UHFFFAOYSA-N
34367
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl
C15H11ClO2
MFCD00001138
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
9H-fluoren-9-ylmethyl carbonochloridate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 249-313-6

RTECSNumber : LQ6250000

TSCA : No

storageNote1 : Moisture Sensitive

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only