missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Fluorenylmethyl Chloroformate 97.0+%, TCI America™

Supplier:  TCI America F019725G

 View more versions of this product

Catalog No. F019725G


Explore available promotions

Description

Description

Specifications

Specifications

9-Fluorenylmethyl Chloroformate [N-Protecting Agent for Peptides Research]
62°C
C15H11ClO2
25 g
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl
258.701
258.70
Crystalline Powder
28920-43-6
White-Yellow
MFCD00001138
3261
IRXSLJNXXZKURP-UHFFFAOYSA-N
9H-fluoren-9-ylmethyl carbonochloridate
34367
≥97.0% (HPLC,T)

Chemical Identifiers

28920-43-6
258.701
IRXSLJNXXZKURP-UHFFFAOYSA-N
34367
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl
C15H11ClO2
MFCD00001138
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
9H-fluoren-9-ylmethyl carbonochloridate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LQ6250000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions