missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-Fluorenylmethyl Chloroformate 97.0+%, TCI America™
Supplier: TCI America F01975G
Specifications
9-Fluorenylmethyl Chloroformate [N-Protecting Agent for Peptides Research] | |
62°C | |
C15H11ClO2 | |
5 g | |
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl | |
258.701 | |
258.70 | |
Crystalline Powder |
28920-43-6 | |
White-Yellow | |
MFCD00001138 | |
3261 | |
IRXSLJNXXZKURP-UHFFFAOYSA-N | |
9H-fluoren-9-ylmethyl carbonochloridate | |
34367 | |
≥97.0% (HPLC,T) |
Chemical Identifiers
28920-43-6 | |
258.701 | |
IRXSLJNXXZKURP-UHFFFAOYSA-N | |
34367 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl |
C15H11ClO2 | |
MFCD00001138 | |
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride | |
9H-fluoren-9-ylmethyl carbonochloridate |
Safety and Handling
RTECSNumber : LQ6250000
TSCA : No
Recommended Storage : Refrigerator