Learn More
9-Phenanthrylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 431730500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 9-Phenanthrylmagnesium bromide | |
| 11.91,82.13 | |
| 0.9450g/mL | |
| AcroSeal™ Glass Bottle | |
| 50mL | |
| 9-phenanthrylmagnesium bromide, 9-phenanthryl-magnesiumbromid, magnesium,bromo-9-phenanthrenyl, magnesium,9h-phenanthren-9-ide,bromide, bromo phenanthren-9-yl magnesium, 9-phenanthrylmagnesium bromide, 0.5 m in thf, 9-phenanthrylmagnesium bromide, 0.25 m in 2-methf, 9-phenanthrylmagnesium bromide 0.5 m in tetrahydrofuran, grignard reagent | |
| RQYSGLNTEAQBIV-UHFFFAOYSA-M | |
| magnesium;9H-phenanthren-9-ide;bromide | |
| 12396678 |
| 71112-64-6,109-99-9 | |
| 17.87,88.09 | |
| −17°C | |
| C14H9BrMg | |
| 0.945 | |
| Solubility in water: reacts. | |
| C1=CC=C2C(=C1)C=[C-]C3=CC=CC=C32.[Mg+2].[Br-] | |
| 281.43 | |
| 281.43 |
Chemical Identifiers
| 71112-64-6 | |
| 281.43 | |
| 9-phenanthrylmagnesium bromide, 9-phenanthryl-magnesiumbromid, magnesium,bromo-9-phenanthrenyl, magnesium,9h-phenanthren-9-ide,bromide, bromo phenanthren-9-yl magnesium, 9-phenanthrylmagnesium bromide, 0.5 m in thf, 9-phenanthrylmagnesium bromide, 0.25 m in 2-methf, 9-phenanthrylmagnesium bromide 0.5 m in tetrahydrofuran, grignard reagent | |
| magnesium;9H-phenanthren-9-ide;bromide |
| C14H9BrMg | |
| RQYSGLNTEAQBIV-UHFFFAOYSA-M | |
| 12396678 | |
| C1=CC=C2C(=C1)C=[C-]C3=CC=CC=C32.[Mg+2].[Br-] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.<br/
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO â Research Use Only