missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pelican™ 9460 Remote Area Lighting System
Convenient and ecologically responsible
$4177.31 - $4177.31
Specifications
Light Source | High Flux LED |
---|---|
Lighting Life | 50,000 hr. |
Voltage | 12 V |
Weight (English) | 55 lb. |
Weight (Metric) | 24.95 kg |
Alert:
The Pelican Products RALS models 9460 and 9470 contain sealed lead acid batteries that can generate hydrogen gas when charging. It is imperative that these products are charged, as instructed, with the case lid open to allow any gases to escape as charging with the lid closed poses a significant hazard. Failure to abide by this instruction could result in injury and/or damage to property.
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
19-502-002
|
Pelican™
9460000110 |
Black |
Each for $4,177.31
|
|
||||||
Description
- Intelligent Control System displays battery life to regulate light intensity and desired run time
- Two swiveling telescope masts and two 360° articulated light arrays for versatile configuration
- Rechargeable battery powers system up to seven hours at peak brightness
- Pelican protector case with pull handle and polyurethane wheels provides durability,easy transport
Specifications
High Flux LED | |
12 V | |
24.95 kg | |
7 hr. | |
CE, RoHs | |
9 in. | |
13.81 in. | |
Remote Area Lighting System |
50,000 hr. | |
55 lb. | |
500 cycles | |
Rechargeable | |
Provides convenient and ecologically responsible alternatives to wasteful generator powered lights | |
22.9 cm | |
35.1 cm |