missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One 96-well Conical Bottom (V) Non-binding Microplates
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Non-binding Microplates is free of detectable DNase, RNase, human DNA.
Supplier: Greiner Bio-One 651901
Description
- Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
- Stable under normal assay conditions without degradation or leaching
- Improves signal-to-noise-ratio
- Manufactured under DIN ISO 9001 guidel
Service Plan
No
Warranty
No
Specifications
No | |
Without Lid | |
Non Binding | |
Polystyrene | |
Non-sterile | |
234 μL | |
Yes | |
40/Cs. | |
Clear |
Standard | |
8 x 12 | |
Clear | |
96 | |
Non-treated | |
Round, Conical Bottom (V-well) | |
10 | |
Yes |