missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One 96-well Conical Bottom (V) Polypropylene Microplates
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.
Supplier: Greiner Bio-One 651205
Description
- Uniform external dimensions
- Well to well spacing of 9mm
- Features alphanumeric coding
- Sealable with adhesive films and heat seal
Specifications
Free of detectable DNase/RNase, human DNA and pyrogens. | |
Without Lid | |
Not Autoclavable | |
Polypropylene | |
Non-sterile | |
100/Cs. |
Standard | |
8 x 12 | |
Green | |
96 | |
Conical Bottom (V-well) | |
128 x 85 x 15 mm |