Thermo Scientific™ 96-Well Flipper™ Microtube Racks
Protect samples and allow racks to be stacked using this 96-well microtube rack.
$118.24 - $118.24
Specifications
Product Type 96-well Flipper Rack
Description Flipper Rack
Material Polypropylene
Stackable Yes
No. per Case 10 Racks
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
21-402-18
View Documents
Thermo Scientific™
8760
Fluorescent Pink
Each for $118.24
Case of 10 Each for $1,182.40
Description
Description
Protect samples and allow racks to be stacked using the Thermo Scientific™ 96-Well Flipper™ Microtube Rack. The rack, which is molded with an alphanumeric grid for easy tube identification, holds 0.5mL tubes on one side and 1.5mL tubes on the other.
  • Holds 0.5mL tubes on one side, 1.5mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Sides have a frosted writing surface for smear-resistant labeling
  • Several colors available
Specifications
Specifications
96-well Flipper Rack
Polypropylene
10 Racks
Flipper Rack
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'