missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One 96-well High Binding Standard ELISA Microplates
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
$222.94 - $887.56
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Binding Type | High Binding |
Color | Clear |
Material | Polystyrene |
Catalog Number | Mfr. No. | Well Shape | Sterility | Well Design | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Well Shape | Sterility | Well Design | Price | Quantity | |||
07000101
|
Greiner Bio-One
655081 |
Flat Bottom (F-well) | Non-sterile | Chimney |
Case of 40 Each for $236.85
|
|
|||
07000102
|
Greiner Bio-One
655061 |
Square, Flat Bottom (F-well) | Non-sterile | Standard |
Case of 40 Each for $242.91
|
|
|||
07000608
|
Greiner Bio-One
650061 |
Square, Round Bottom (U-well) | Non-sterile | Round |
Case of 40 Each for $222.94
|
|
|||
07000995
|
Greiner Bio-One
65006125 |
Round Bottom (U-well) | Sterile | Standard |
Case of 150 Each for $887.56
|
|
|||
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Specifically tre
Specifications
Standard | |
High Binding | |
Polystyrene | |
Microlon 600 |
8 x 12 | |
Clear | |
96 |