missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ 96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Supplier: Thermo Scientific™ 9205
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
- For high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Standard | |
8 x 12 | |
Microtiter | |
Microplate | |
330 μL | |
Non-sterile | |
C | |
10 |
Without Lid | |
50 Cases, 10 Pk. | |
Clear | |
Polystyrene | |
96 | |
Non-treated | |
50 | |
50/Pk. |