Learn More
Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats
Save time and minimize the risk of cross-contamination with convenient 96-Well Plate Kits packaged with pre-assembled glass or PTFE vials.
$199.49 - $973.10
Specifications
Format Standard
Array 8 x 12
Product Line Webseal
Product Type Glass Inserted Plate Kit with 96-Position Mat
No. of Wells 96
View More Specs
Products 5
Catalog Number Mfr. No. Total Volume (Metric) Well Profile Includes Price Quantity  
Catalog Number Mfr. No. Total Volume (Metric) Well Profile Includes Price Quantity  
03-252-191
View Documents
Thermo Scientific™
60180K100
500 μL Taper Vial, Mat
Pack of 5 Each for $931.81
Only null left
03-252-192
View Documents
Thermo Scientific™
60180K101
700 μL Taper Vial, Mat, Cutting Tool
Pack of 5 Each for $792.19
Only null left
03-252-194
View Documents
Thermo Scientific™
60180K104
1100 μL Round Vial, Mat, Cutting Tool
Pack of 5 Each for $792.19
Only null left
03-252-196
View Documents
Thermo Scientific™
60180K106
1500 μL Round Vial, Mat
Pack of 5 Each for $973.10
Only null left
03-252-197
View Documents
Thermo Scientific™
60180K107
1000 μL Constricted Vial, Mat
Pack for $199.49
Only null left
Description
Description
Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.
  • Convenient wellplate kits include pre-inserted glass or PTFE vials and 96-position sealing mats; some include cutting tool
  • Mats seal samples for storage
  • Plates are glass-inserted for greater inertness compared to pure PP plates
  • Unique cutting tool allows removal of individual samples with the seal in place, eliminating cross-contamination
Specifications
Specifications
Standard
Webseal
96
8 x 12
Glass Inserted Plate Kit with 96-Position Mat
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.