Learn More
Thermo Scientific™ WebSeal 96-Well Non-Coated Plastic Microplates
For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 96-well microplates.
$669.32 - $1805.44
Spécifications
Format | Standard |
---|---|
Array | 8 x 12 |
Product Line | Webseal |
Product Type | Microplate |
Material | Polypropylene |
Numéro de catalogue | Numéro du manufacturier. | Total Volume (Metric) Well | Well Shape | No. per Pack | Prix | Quantité | |||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Total Volume (Metric) Well | Well Shape | No. per Pack | Prix | Quantité | |||
13-622-482
|
Thermo Scientific™
60180P129 |
145 μL | Conical Bottom (V-well) | 100 Microplates |
paquet de 100 chaque for $1,472.25
|
|
|||
13-622-481
|
Thermo Scientific™
60180P127 |
252 μL | Conical Bottom (V-well) | 100 Microplates |
paquet de 100 chaque for $1,784.83
|
|
|||
60180P154
|
Thermo Scientific™
60180P154 |
1100 μL | Round Bottom (U-well) | 50 Microplates |
paquet de 50 chaque for $762.11
|
|
|||
13-622-480
|
Thermo Scientific™
60180P124 |
2000 μL | Round Bottom (U-well) | 50 Microplates |
paquet de 50 chaque for $669.32
|
|
|||
03-252-407
|
Thermo Scientific™
60180P134 |
2000 μL | Round Bottom (U-well) | 60 Microplates |
caisse de 60 chaque for $1,805.44
|
|
|||
Description
Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 96-Well Non-Coated Plastic Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:
- Pharmaceutical and industrial QA/QC
- High-throughput screening (HTS)
- Sample collection and storage
- Combinatorial chemistry
- Liquid phase assay
Chemically and Thermally Resistant Polypropylene
- Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
- Will tolerate temperatures from −80°C to 121°C
- GC-tested to ensure low extractables
Broad Instrument Compatibility
- SBS and ANSI standard footprint design
Well Formats to Suit Every Application
- Shallow well and deep well formats
- U- and V- bottom wells ensure optimal sample recovery
Spécifications
Standard | |
Webseal | |
Polypropylene | |
Non-coated |
8 x 12 | |
Microplate | |
Non-sterile |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.