missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BRAND™ 96-well Plates
Excellent for cell and tissue culture, sample storage, immunological assays and other applications
$892.19 - $892.19
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Color | Clear |
Product Type | 96-well Assay Microplate |
Material | Polypropylene |
Catalog Number | Mfr. No. | Material | Product Type | Volume (Metric) Well | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Product Type | Volume (Metric) Well | Price | Quantity | |||
13-882-92
|
BRAND™
701330 |
Polypropylene | 96-well Assay Microplate | 330 μL |
Pack of 100 Each for $892.19
|
|
|||
Description
- Polypropylene plates are autoclavable at 250°F (121°C) and resistant to many chemicals, including phenols, chloroform and DMSO.
- Clear polystyrene plates are available in non-sterile, sterile, and sterile with treated hydrophilic surfaces, in your choice of flat, round, or conical bottoms.
- Plates have raised rims to simplify sealing with adhesive films, and recessed interwell spaces to reduce well cross-contamination. Optional lids have condensation rings.
Specifications
Standard | |
Clear | |
Polypropylene | |
96 | |
Round |
8 x 12 | |
96-well Assay Microplate | |
330 μL | |
Non-sterile | |
Cell and tissue culture, sample storage, immunological assays and other applications |