Learn More
Thallous Acetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 420775000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Thallous acetate | |
500 g | |
C2H3O2Tl | |
MFCD00013045 | |
thallium i acetate | |
[HH].CC(=O)O.[Tl] | |
263.42 | |
263.42 | |
Authentic |
563-68-8 | |
99% | |
CH3COOTl | |
15, 9405 | |
CNWGLQAFFSLHRX-UHFFFAOYSA-N | |
acetic acid;molecular hydrogen;thallium | |
131675083 | |
99% | |
Glass bottle |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-257-5
RUO â Research Use Only